Learn More
Cayman Chemical Immunoassay Buffer E Concentrate (10X), 10 mL
SDP
Catalog No. 502893387
Encompass_Preferred
Immunoassay Buffer E Concentrate (10X) has been tested and formulated to work exclusively with Cayman’s MHC Class I (human) ELISA Kit (Item No.502060).
Catalog No. 50-289-3387 Supplier Cayman Chemical Supplier No. 50206410 ML
Immunoassay Buffer E Concentrate (10X) has been tested and formulated to work exclusively with Cayman’s MHC Class I (human) ELISA Kit (Item No.502060).