Learn More
Cayman Chemical ImunoOncology Screeng LIb 25uL
SDP
Catalog No. 502938748
Encompass_Preferred
An antibiotic active against Gram positive and Gram negative bacteria at 255 Ug/ml inhibits bacterial protein synthesis (EC50 0 4 UM in E coli) increases EF-Tu-associated GTPase activity (EC50 6 UM in E coli)
Catalog No. 50-293-8748 Supplier Cayman Chemical Supplier No. 2876025 UL
An antibiotic active against Gram positive and Gram negative bacteria at 255 Ug/ml inhibits bacterial protein synthesis (EC50 0 4 UM in E coli) increases EF-Tu-associated GTPase activity (EC50 6 UM in E coli)