Learn More
Cayman Chemical ImunoOncology Screeng LIb 50uL
SDP
Catalog No. 502937618
Encompass_Preferred
An antibiotic active against Gram positive and Gram negative bacteria at 255 Ug/ml inhibits bacterial protein synthesis (EC50 0 4 UM in E coli) increases EF-Tu-associated GTPase activity (EC50 6 UM in E coli)
Catalog No. 50-293-7618 Supplier Cayman Chemical Supplier No. 2876050 UL
An antibiotic active against Gram positive and Gram negative bacteria at 255 Ug/ml inhibits bacterial protein synthesis (EC50 0 4 UM in E coli) increases EF-Tu-associated GTPase activity (EC50 6 UM in E coli)