Learn More
Takara Bio In-Fusion HD Cloning Kit | 100 Rxns
SDP
Catalog No. NC1454739
Encompass
The In-Fusion HD Cloning Kit enables high-efficiency, high-fidelity, directional cloning of one or more PCR fragments into any vector. This package includes 5X In-Fusion HD Enzyme Premix for 100 reactions, linearized pUC19 Control Vector, and a 2-kb control insert.
Catalog No. NC1454739 Supplier Takara Bio Supplier No. 639650
The In-Fusion HD Cloning Kit enables high-efficiency, high-fidelity, directional cloning of one or more PCR fragments into any vector. This package includes 5X In-Fusion HD Enzyme Premix for 100 reactions, linearized pUC19 Control Vector, and a 2-kb control insert.