Learn More
BOSTON FLUID SYSTEMS INC IN-LINE PARTICULATE FILTER
Supplier:  BOSTON FLUID SYSTEMS INC SS2F05
Encompass
Stainless Steel In-Line Particulate Filter, 1/8 in. Swagelok Tube Fitting, 0.5 Micron Pore Size
Catalog No. NC1473415