Learn More
Grainger Zoro Select In Line Post, Steel, 96 in., Yellow, 14 ga
Catalog No. 502779253
Encompass_Preferred
This line post is designed for wire partition systems, providing structural support with a durable finish. Constructed from steel, it is finished with a powder-coated yellow color to ensure visibility and resistance to wear.
  • Meets or exceeds OSHA and ANSI specifications
  • 14 ga steel construction for durability
  • Includes mounting hardware such as 1/2 in. anchor bolts and brackets with self-tapping screws
  • Features a 6 in. x 6 in. footplate with six 5/8 in. diameter mounting holes
  • Designed as a line post for wire partition configurations
  • Overall height of 8 ft. with a 2 in. x 2 in. post profile
Catalog No. 50-277-9253 Supplier Grainger Supplier No. 19N874
This line post is designed for wire partition systems, providing structural support with a durable finish. Constructed from steel, it is finished with a powder-coated yellow color to ensure visibility and resistance to wear.
  • Meets or exceeds OSHA and ANSI specifications
  • 14 ga steel construction for durability
  • Includes mounting hardware such as 1/2 in. anchor bolts and brackets with self-tapping screws
  • Features a 6 in. x 6 in. footplate with six 5/8 in. diameter mounting holes
  • Designed as a line post for wire partition configurations
  • Overall height of 8 ft. with a 2 in. x 2 in. post profile