Learn More
MilliporeSigma™ Supelco™ In-Line Pressure Regulator, Economy Model
Catalog No. 111010231 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Regulator Body, Gauge, Panel Mount Bracket
Catalog No. Description
11-101-0231 Regulator Body, Gauge, Panel Mount Bracket
1 options
Catalog No. 11-101-0231 Supplier MilliporeSigma™ Supelco™ Supplier No. 23831U
Add to Cart
Add to Cart
An inexpensive version for general lab applications
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Pneumatic Control, and Other Non-Chromatographic Purposes
For Use With (Equipment) Gas Streams
Includes 1/8 in. Brass Swagelok Nuts and Ferrules
Model Economy
Pressure 0 to 300 psi (Inlet), 0 to 50 psi (Outlet)
Quantity 1 Ea.
Description Regulator Body, Gauge, Panel Mount Bracket