Learn More
MilliporeSigma™ Supelco™ In-Line Pressure Regulators
Catalog No. p-7213644 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Ultra-High Purity Version
Catalog No. Description
11-100-3754 Ultra-High Purity Version
1 options
Catalog No. 11-100-3754 Supplier MilliporeSigma™ Supelco™ Supplier No. 23884
Only null left
Add to Cart
Add to Cart
Install downstream of a gas cylinder regulator to adjust pressure closer to the point of use
  • Adjusts pressure closer to the point of use
  • Includes 1/8 in. Swagelok Nuts and Ferrules
  • Maximum Input Pressure: 400 psi
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Air, H2, N2, Ar, He
Includes 1/8 in. Swagelok Nuts and Ferrules
Pressure 0-100 psi (Outlet)
Description Ultra-High Purity Version
Type In-Line Pressure Regulator