Learn More
Alkali Scientific ASI™ In Line Vacuum Filter, PES
SDP
Supplier:  Alkali Scientific TE50S
Encompass_Preferred
In-Line Vacuum Filter Inlet and Outlet for Silicone Tubing Protect vacuum sources from contamination and moisture damage STERILE are individually wrapped and 25/Pk Membranes made out of Polyethersulfone or Polyvinylidenflouride, Housing 50mm, Pore 0.22µm.
Catalog No. 50-302-8889