Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ In Search of the Cholesterol Gene
SDPSCI_ED

Catalog No. S66830 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S66830 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66830 Supplier Edvotek™ Supplier No. 316
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, cholesterol standard solution, standard DNA markers, control samples, simulated patient serum samples and DNA samples, cholesterol oxidase enzyme, potassium iodide, color enhancer & color developer, agarose, electrophoresis buffer, InstaStain™ Ethidium Bromide

Requires

  • Horizontal gel electrophoresis apparatus
  • Hotplate, micropipets, balance, waterbath, and spectrophotometer
  • 250mL Flasks
  • UV transilluminator
  • All necessary safety equipment
  • Distilled or deionized water

This experiment includes reagents for the colorimetric enzymatic reaction which is the basis of the clinical cholesterol test. In addition, using agarose gel electrophoresis, students will analyze a simulated genetic screening for a disease.

Coronary heart disease and stroke are major causes of death in the Western world. Elevated blood cholesterol levels are a serious risk factor for both conditions. The genetic disease familial hypercholesterolemia (FH) causes an increase in blood levels of the 'bad' form of cholesterol, low-density lipoprotein (LDL). In untreated patients with the mutant FH gene, the condition can cause premature death. This experiment includes reagents for the colorimetric enzymatic reaction which is the basis of the clinical cholesterol test. In addition, using agarose gel electrophoresis, students will analyze a simulated genetic screening for a disease.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Agarose gel electrophoresis
Includes Instructions, cholesterol standard solution, standard DNA markers, control samples, simulated patient serum samples and DNA samples, cholesterol oxidase enzyme, potassium iodide, color enhancer and color developer, agarose, electrophoresis buffer, InstaStain™ ethidium bromide
Requires Horizontal gel electrophoresis apparatus, power supply, automatic micropipette with tips, balance, microwave oven or hot plate, incubation oven or water bath, spectrophotometer and cuvettes, UV transilluminator, pipet pumps or bulbs, lab glassware, large weigh boats
Time Range Complete in 2 hours 15 minutes
Content And Storage Some components require freezer and refrigerator storage
Class Size 10 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.