Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ In Search of the Kissing Disease
SDPSCI_ED

Catalog No. S64844 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S64844 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S64844 Supplier Edvotek™ Supplier No. 274
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, samples, antigens; antibodies, various solutions and reagents, pipets and microtest tubes

Infectious mononucleosis is common among young people of high school and college age.

  • The causative agent is Epstein-Barr virus (EBV) which can be passed on through saliva during kissing
  • Search for the presence of EBV using the ELISA reaction to detect specific viral proteins
  • No human or viral materials used in this experiment

Specifications

Grade 9 to Undergraduate
For Use With (Application) Perform an enzyme-linked immunosorbent assay (ELISA) in order to screen simulated serum samples for antibodies to the Epstein Barr Virus (EBV)
Includes Instructions, 10X ELISA wash buffer, ELISA dilution buffer, antigen (lyophilized), primary antibody (lyophilized), secondary antibody (lyophilized), ABTS substrate (lyophilized), and ABTS reaction buffer, small transfer pipets, microplate, 15 mL conical tubes, and snap-top microcentrifuge tubes
Requires Automatic micropipettes with tips (recommended), paper towels, distilled or deionized water, beakers or flasks, disposable lab gloves, and safety goggles
Time Range 50 min.
Content And Storage Refrigerator
Class Size 10 Lab Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.