Learn More
Mitegen In Situ 1 Crystallization Plate, For Direct X-ray Diffraction Analysis, Minimizes Sample Handling, 100 Plate Pack
SDP
Catalog No. 5000495018
Encompass_Preferred
The In Situ 1 Crystallization Plate is specifically designed to streamline the protein crystallization workflow, enabling direct X-ray diffraction analysis without the need for traditional crystal harvesting and mounting. This innovative plate format significantly minimizes sample handling, reducing the risk of crystal damage or contamination during sensitive experiments. It provides an optimized environment for crystal growth and subsequent direct observation, making it ideal for high-throughput screening and detailed structural biology studies. Manufactured from materials chosen for their optical clarity and minimal X-ray background interference, these plates ensure high-quality imaging and reliable diffraction data. Their design facilitates efficient sample visualization and alignment, enhancing experimental reproducibility. This is a pack of 100 plates.
Catalog No. 50-004-95018 Supplier Mitegen Supplier No. INSITU01CL100
The In Situ 1 Crystallization Plate is specifically designed to streamline the protein crystallization workflow, enabling direct X-ray diffraction analysis without the need for traditional crystal harvesting and mounting. This innovative plate format significantly minimizes sample handling, reducing the risk of crystal damage or contamination during sensitive experiments. It provides an optimized environment for crystal growth and subsequent direct observation, making it ideal for high-throughput screening and detailed structural biology studies. Manufactured from materials chosen for their optical clarity and minimal X-ray background interference, these plates ensure high-quality imaging and reliable diffraction data. Their design facilitates efficient sample visualization and alignment, enhancing experimental reproducibility. This is a pack of 100 plates.