Learn More
Alkali Scientific In-Situ Adapter for Use with Combination Blocks
SDP
Catalog No. 502389169
Encompass_Preferred
InSitu adapter fits on the TC 9639 Thermal Cycler block and holds 4 standard slides. Compatible with: T5000-96 and T5000-384.
Catalog No. 50-238-9169 Supplier Alkali Scientific Supplier No. T5000AIS
InSitu adapter fits on the TC 9639 Thermal Cycler block and holds 4 standard slides. Compatible with: T5000-96 and T5000-384.