Learn More
Enzo Life Sciences In situ hybridization buffer (1.25X concentrate) (10 ml)
SDP
Catalog No. 501031805
Encompass_Preferred
In Situ Hybridization Buffer provides a high quality hybridization medium for in situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, digoxigenin- or fluorescein-modified nucleotides. Formulation: Liquid. Sodium phosphate, NaCl and EDTA (SSPE) buffered formamide, containing hybridization enhancers. Shipping: Shipped on Blue Ice. Long Term Storage: +4°C
Catalog No. 50-103-1805 Supplier Enzo Life Sciences Supplier No. ENZ33808
In Situ Hybridization Buffer provides a high quality hybridization medium for in situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, digoxigenin- or fluorescein-modified nucleotides. Formulation: Liquid. Sodium phosphate, NaCl and EDTA (SSPE) buffered formamide, containing hybridization enhancers. Shipping: Shipped on Blue Ice. Long Term Storage: +4°C