Learn More
Electron Microscopy Sciences In-Situ Mount 30 ML
SDP
Catalog No. 5030246
Encompass_Preferred
In-Situ mounting medium is a unique mounting medium designed for permanent preservation of tissue sections and cell smear with alkaline phosphatase chromogens like NBT/BCIP and red stain. Use for mounting tissues and cell smears.
Catalog No. 50-302-46 Supplier Electron Microscopy Sciences Supplier No. 1798830
In-Situ mounting medium is a unique mounting medium designed for permanent preservation of tissue sections and cell smear with alkaline phosphatase chromogens like NBT/BCIP and red stain. Use for mounting tissues and cell smears.