Learn More
Bioworld In Situ™ Tissue Fixative, 500 mL
Catalog No. 50199056
Encompass_Preferred
In SituT Fixative is utilized for routine, manual or automated tissue fixation. Cellular material or tissues can be immersed in In Situ for transport and preservation of antigenic sites. In Situ maintains the morphological integrity of cells or tissues, and enhances specific immunohistochermical staining in tissues embedded in paraffin medium.• In Situ permits in situ PCR reactions.• A nonhazardous tissue fixative exempt from EPA disposal regulations.• Virtually odorless.• Inactivates the HIV-1 virus in culture.• Maintains cellular morphology.• Optimizes immunohistochemistry results.
Catalog No. 50-199-056 Supplier Bioworld Supplier No. 217300102
In SituT Fixative is utilized for routine, manual or automated tissue fixation. Cellular material or tissues can be immersed in In Situ for transport and preservation of antigenic sites. In Situ maintains the morphological integrity of cells or tissues, and enhances specific immunohistochermical staining in tissues embedded in paraffin medium.• In Situ permits in situ PCR reactions.• A nonhazardous tissue fixative exempt from EPA disposal regulations.• Virtually odorless.• Inactivates the HIV-1 virus in culture.• Maintains cellular morphology.• Optimizes immunohistochemistry results.