Learn More
MSC Betco Dishwashing Detergent, Concentrate, 1 gal, Fresh Ozonic Scent, NSF Certified
Catalog No. 502836013
Encompass
Betco Symplicity In-Sync Hand Dishwashing Detergent is a concentrated liquid designed for commercial kitchens and food service environments. This premium detergent effectively cuts through tough grease and baked-on food, leaving dishes sparkling clean and film-free. It's formulated to be mild on hands when diluted and ensures fast grease emulsification with lasting suds.
  • Improved formula for proper dispensing equipment.
  • Leaves dishes sparkling clean and film-free.
  • Quickly removes baked-on food and grease from pots and pans.
  • Rinses freely and never streaks.
  • Concentrated formula uses 1 to 2 oz. per 10 gallons of water.
  • Lasting suds and fast grease emulsification.
  • Mild and gentle to hands when diluted.
  • Certified for regulated food processing and food service environments.
Catalog No. 50-283-6013 Supplier MSC Supplier No. 73011264
Betco Symplicity In-Sync Hand Dishwashing Detergent is a concentrated liquid designed for commercial kitchens and food service environments. This premium detergent effectively cuts through tough grease and baked-on food, leaving dishes sparkling clean and film-free. It's formulated to be mild on hands when diluted and ensures fast grease emulsification with lasting suds.
  • Improved formula for proper dispensing equipment.
  • Leaves dishes sparkling clean and film-free.
  • Quickly removes baked-on food and grease from pots and pans.
  • Rinses freely and never streaks.
  • Concentrated formula uses 1 to 2 oz. per 10 gallons of water.
  • Lasting suds and fast grease emulsification.
  • Mild and gentle to hands when diluted.
  • Certified for regulated food processing and food service environments.