Learn More
Cell Biolabs Inc OxiSelect™ In Vitro ROS/RNS Assay Kit (Green Fluorescence), Trial Size, 20 assays
Catalog No. NC0729674
Encompass_Preferred
The OxiSelect™ In Vitro ROS/RNS Assay Kit is an assay for measuring the total free radical presence of a sample. The assay employs a proprietary quenched fluorogenic probe, DCFH-DiOxyQ), which is based on similar chemistry to the popular DCFH-DA. The DCFH-DiOxyQ probe is first primed with a quench removal reagent, and subsequently stabilized in the highly reactive DCFH form. In this reactive state, ROS and RNS species can react with DCFH, which is rapidly oxidized to the highly fluorescent 2’, 7’-dichlorodihydrofluorescein (DCF) . The DCFH-DiOxyQ probe can react with hydrogen peroxide, peroxyl radical, nitric oxide, and peroxynitrite anion. These free radical molecules are representative of both ROS and RNS, thus allowing for measurement of the total free radical population within a sample. The kit has a detection sensitivity limit of 40 nM for H2O2. Each trial size kit provides sufficient reagents to perform up to 20 assays, including standard curve and unknown samples.
Catalog No. NC0729674 Supplier Cell Biolabs Inc Supplier No. STA347T
The OxiSelect™ In Vitro ROS/RNS Assay Kit is an assay for measuring the total free radical presence of a sample. The assay employs a proprietary quenched fluorogenic probe, DCFH-DiOxyQ), which is based on similar chemistry to the popular DCFH-DA. The DCFH-DiOxyQ probe is first primed with a quench removal reagent, and subsequently stabilized in the highly reactive DCFH form. In this reactive state, ROS and RNS species can react with DCFH, which is rapidly oxidized to the highly fluorescent 2’, 7’-dichlorodihydrofluorescein (DCF) . The DCFH-DiOxyQ probe can react with hydrogen peroxide, peroxyl radical, nitric oxide, and peroxynitrite anion. These free radical molecules are representative of both ROS and RNS, thus allowing for measurement of the total free radical population within a sample. The kit has a detection sensitivity limit of 40 nM for H2O2. Each trial size kit provides sufficient reagents to perform up to 20 assays, including standard curve and unknown samples.