Learn More
Cayman Chemical In vitro Screening for Drug-Induced Hepatotoxicity using upcyte Hepatocytes Hepatotoxicity Assay, 96 Well
SDP
Catalog No. 502988225
Encompass_Preferred
The reagents and materials provided in this assay kit are formatted to allow the user to choose between two alternative assay setups. In one scenario 48 culture wells may be setup at two different times. In the other assay scenario 96 culture wells may be setup at one time. This kit includes two aliquots of cryopreserved Luminescent Reporter Hepatocytes (upcyte), donor 10-13, isolated from an adult Caucasian female, that have been further modified to constitutively express the luciferase enzyme. The hepatocytes are prepared using INDIGO's proprietary CryoMite™ process, which yields high cell viability post-thaw, and provides the convenience of immediately dispensing cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. An overnight culture period allows full recovery of the post-thaw hepatocytes and.
Catalog No. 50-298-8225 Supplier Cayman Chemical Supplier No. 3026096 WELL
The reagents and materials provided in this assay kit are formatted to allow the user to choose between two alternative assay setups. In one scenario 48 culture wells may be setup at two different times. In the other assay scenario 96 culture wells may be setup at one time. This kit includes two aliquots of cryopreserved Luminescent Reporter Hepatocytes (upcyte), donor 10-13, isolated from an adult Caucasian female, that have been further modified to constitutively express the luciferase enzyme. The hepatocytes are prepared using INDIGO's proprietary CryoMite™ process, which yields high cell viability post-thaw, and provides the convenience of immediately dispensing cells into assay plates. There is no need for intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. An overnight culture period allows full recovery of the post-thaw hepatocytes and.