Learn More
Sigma Aldrich Fine Chemicals Biosciences In Vitro Toxicology Assay
Catalog No. 502901932
Encompass
Tox7 is a member of Sigmas CytoToxicity Kit series. It is based on the measurement of lactate dehydrogenase (LDH) release from cells as a measure of cell membrane damage and cytotoxicity. This technique has been utilized as an alternative to 51Cr release for cell mediated cytotoxicity assays as well as conventional cytotoxicity resulting from interaction of a test material with the cell.
Catalog No. 50-290-1932 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TOX71KT
Tox7 is a member of Sigmas CytoToxicity Kit series. It is based on the measurement of lactate dehydrogenase (LDH) release from cells as a measure of cell membrane damage and cytotoxicity. This technique has been utilized as an alternative to 51Cr release for cell mediated cytotoxicity assays as well as conventional cytotoxicity resulting from interaction of a test material with the cell.