Learn More
Sigma Aldrich In Vivo Edu Flow Cytometry Kit 594
Catalog No. BCK594IVFCM Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Catalog No. BCK594IVFCM Supplier Sigma Aldrich Supplier No. BCK594IVFCM
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The Baseclick EdU In Vivo Kits provide a superior alternative to BrdU and [3H]thymidine assays for detection of cell proliferation in whole animals, showing no toxic effects. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog to thymidine and is incorporated into DNA during active DNA synthesis. In contrast to BrdU assays, the EdU In Vivo Assays are not antibody based and therefore do not require DNA denaturation for detection of the incorporated nucleoside. Instead, the EdU In Vivo Kits utilize click chemistry for detection in a variety of dye fluorescent readouts. Furthermore, the streamlined detection protocol reduces both the total number of steps and significantly decreases the total amount of time. The simple click chemistry detection procedure is complete within 30 minutes and is compatible with multiplexing for content and context-rich results.The in vivo kits are designed in a sense that you combine one of the three baseclick EdU cell proliferation kits such as:1. Imaging kit (BCK-IV-IM)2. Flow cytometry kit (BCK-IV-FC)3. High throughput screening – HTS kit (BCK-IV-HTS) You can select the above kits with the dye of choice and the right EdU content for your animal model. Depending on your animal model or the number of animals to be tested you can choose between three kit sizes S, M, and L with increasing EdU content, as indicated in the following table. Kit Size Content of EdU S 50 mg M 500 mg L 1000 mgin Vivo, Flow Cytometry