Learn More
Cambridge Isotope Laboratories L-METHIONINE (2,3,3,4,4-D5, 98%; METHYL-13CH3, 99%), 0.5 G
Catalog No. 501629821
Encompass
L-METHIONINE (2,3,3,4,4-D5, 98%; METHYL-13CH3, 99%), 0.5 G
Catalog No. 50-162-9821 Supplier Cambridge Isotope Laboratories Supplier No. CDLM88850.5
L-METHIONINE (2,3,3,4,4-D5, 98%; METHYL-13CH3, 99%), 0.5 G