Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Litron Laboratories IN VIVO MICROFLOW KIT

Catalog No. NC3717991
Encompass

  • Kit type: In Vivo MicroFlow
  • Target species: Rat
  • Sample type: Whole Blood
  • Samples per kit: 60 samples
  • Analysis service: Included
  • Blood collection tubes: K2EDTA Vacutainer Tubes
  • Anticoagulant: K2EDTA
  • Diluent: Anticoagulant/Diluent provided
  • Shipping container: Exakt-Pak Shipping Container
  • Temperature control: Cold Packs included
  • Assay principle: Flow Cytometry
  • Endpoint measured: Micronuclei, Mature and Immature Red Blood Cells (%RET)
  • Key antibody: Anti-CD71
  • Micronucleus detection: DNA stain
  • Reproducibility: Calibration Standards provided
  • Application: DNA Damage Assessment, In Vivo Micronucleus Assay

Catalog No. NC3717991 Supplier Litron Laboratories Supplier No. BASICR WHOLE BLOOD
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

  • Kit type: In Vivo MicroFlow
  • Target species: Rat
  • Sample type: Whole Blood
  • Samples per kit: 60 samples
  • Analysis service: Included
  • Blood collection tubes: K2EDTA Vacutainer Tubes
  • Anticoagulant: K2EDTA
  • Diluent: Anticoagulant/Diluent provided
  • Shipping container: Exakt-Pak Shipping Container
  • Temperature control: Cold Packs included
  • Assay principle: Flow Cytometry
  • Endpoint measured: Micronuclei, Mature and Immature Red Blood Cells (%RET)
  • Key antibody: Anti-CD71
  • Micronucleus detection: DNA stain
  • Reproducibility: Calibration Standards provided
  • Application: DNA Damage Assessment, In Vivo Micronucleus Assay

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.