Learn More
Litron Laboratories MicroFlow Archive Stage 1, Rat Kit
Catalog No. NC1261758
Encompass
Each MicroFlow Archive Stage 1, Rat kit contains all of the quality-assured supplies & reagents to collect, fix and store 60 rat blood samples at ultracold temperatures. Note: analysis of samples requires a Stage 2 kit (ordered separately).
Catalog No. NC1261758 Supplier Litron Laboratories Supplier No. STAGE 1,RAT
Each MicroFlow Archive Stage 1, Rat kit contains all of the quality-assured supplies & reagents to collect, fix and store 60 rat blood samples at ultracold temperatures. Note: analysis of samples requires a Stage 2 kit (ordered separately).