Learn More
Cytek Biosciences Inc Antibodies In Vivo Ready™ Anti-Mouse IFN gamma (XMG1.2) ..... Cat. No. 40-7311-U500 ..... Size 500 µg
Supplier:  Cytek Biosciences Inc 407311U500
Encompass_Preferred
Antibodies In Vivo Ready™ Anti-Mouse IFN gamma (XMG1.2) ..... Cat. No. 40-7311-U500 ..... Size 500 µgThe XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20 kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells which allow the cytokine to modulate innate immunity when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein, e.g. by flow cytometry, as well as for quantitative analysis of the secreted protein by ELISA when paired with an appropriate capture antibody. This clone is also widely used for neutralization of the functional activity of IFN-g in a variety of assays.
Catalog No. 50-210-3294