Learn More
Microbiologics™ Inactivated Chlamydia pneumoniae
SDP
Catalog No. 23047390 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
23-047-390 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-047-390 Supplier Microbiologics™ Supplier No. HE0034N
Add to Cart
Add to Cart
This item is not returnable. View return policy
Inactivated Helix Elite Molecular Standards designed, full process controls for use in molecular testing
  • Uses:
    • For use in molecular testing
  • Includes:
    • 5 unassayed lyophilized pellets (individual vials to avoid cross contamination)
Specifications
Biosafety Level 1
Content And Storage 2 to 25°C
Manufacturing Origin United States
Product Type Microbiology Media
No. per Pack 5 Pellets/Kit
Packaging Type Plastic box
Quantity 1 Kit