Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Inactivated Influenza A/B and Respiratory Syncytial Virus
SDP

Catalog No. 23047302 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-302 5 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-302 Supplier Microbiologics™ Supplier No. HE0044N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inactivated Helix Elite Molecular Standards are used for every step of the molecular testing process. Quick dissolving lyophilized pellets may be reconstituted in buffer or transport medium. These are FDA listed and CE Marked as an In-Vitro Diagnostic (IVD) Medical Device.

  • Inactivated Helix Elite Molecular Standards are designed for use in molecular testing.
  • Each kit contains 5 unassayed lyophilized pellets, packaged in individual vials for ease-of-use and to avoid cross contamination.
  • For in vitro diagnostic use
  • Biosafety Level: 1
  • Each of the 5 pellets contains:
  • Influenza A (H1N1) Virus, Influenza B Virus, Strain Hong Kong/5/72, Respiratory Syncytial Virus (RSV) A, Strain Long

Specifications

Biosafety Level 1
Content And Storage 2 to 25°C
Format Helix Elite Inactivated Standard
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) Molecular Testing
Packaging Type Plastic Box
Quantity 5 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.