Learn More
Aviva Systems Biology Corp PATJ Antibody - N-terminal region (ARP42938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INADL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human INADL. Alternate names: Cipp, FLJ26982, PATJ, hINADL, InaD-likeApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58368