Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inarigivir ammonium | 2172788-92-8 | 99.0% | 604.53 g·mol⁻¹ | C20H29N8O10PS | 1 ML
SDP

Catalog No. 5000247965
Encompass_Preferred

Inarigivir ammonium is a dinucleotide antiviral compound reported to inhibit hepatitis B virus (HBV). It is supplied for research use in both solid form and as a ready-to-use 10 mM solution in DMSO (1 mL), and is characterized by high purity with defined storage recommendations.

  • Dinucleotide antiviral with reported HBV inhibitory activity.
  • Available as solid or 10 mM solution in DMSO for flexible use.
  • High reported purity (approximately 99.0%).
  • Molecular weight 604.53 g·mol⁻¹; formula C20H29N8O10PS.
  • Solid storage: sealed at 4°C, away from moisture.
  • Solution storage: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-47965 Supplier Medchemexpress LLC Supplier No. HY101954A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inarigivir ammonium is a dinucleotide antiviral compound reported to inhibit hepatitis B virus (HBV). It is supplied for research use in both solid form and as a ready-to-use 10 mM solution in DMSO (1 mL), and is characterized by high purity with defined storage recommendations.

  • Dinucleotide antiviral with reported HBV inhibitory activity.
  • Available as solid or 10 mM solution in DMSO for flexible use.
  • High reported purity (approximately 99.0%).
  • Molecular weight 604.53 g·mol⁻¹; formula C20H29N8O10PS.
  • Solid storage: sealed at 4°C, away from moisture.
  • Solution storage: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.