Learn More
Medchemexpress LLC Inarigivir ammonium | 2172788-92-8 | 99.0% | 604.53 g/mol | C20H29N8O10PS | 5 MG
SDP
Catalog No. 5000246572
Encompass_Preferred
Inarigivir ammonium is the ammonium salt of a dinucleotide antiviral developed as an HBV inhibitor for preclinical research. It reduces liver HBV DNA in animal models and is supplied with defined solubility and storage recommendations for laboratory use.
  • Dinucleotide antiviral active against hepatitis B virus.
  • Provided as an ammonium salt suitable for research use.
  • High purity (99.0%) for consistent experimental results.
  • Soluble in DMSO, water, and PBS at high concentrations; ultrasonic assistance recommended.
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, including 5 mg packages.
Catalog No. 50-002-46572 Supplier Medchemexpress LLC Supplier No. HY101954A5MG
Inarigivir ammonium is the ammonium salt of a dinucleotide antiviral developed as an HBV inhibitor for preclinical research. It reduces liver HBV DNA in animal models and is supplied with defined solubility and storage recommendations for laboratory use.
  • Dinucleotide antiviral active against hepatitis B virus.
  • Provided as an ammonium salt suitable for research use.
  • High purity (99.0%) for consistent experimental results.
  • Soluble in DMSO, water, and PBS at high concentrations; ultrasonic assistance recommended.
  • Stable under recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, including 5 mg packages.