Learn More
Medchemexpress LLC Inarigivir ammonium | 2172788-92-8 | 99.0% | 604.53 g·mol⁻¹ | C20H29N8O10PS | 50 MG
SDP
Catalog No. 5000249119
Encompass_Preferred
Inarigivir ammonium is a dinucleotide research compound that functions as a RIG-I agonist and has been reported to reduce hepatitis B virus (HBV) DNA in preclinical models. It is provided for laboratory research in solid form and as DMSO solutions, and is characterized by high reported purity and defined physicochemical data.
  • Acts as a RIG-I agonist to activate innate immune responses.
  • Demonstrated antiviral activity against hepatitis B virus in preclinical studies.
  • Supplied as a solid and as 10 mM DMSO solutions for flexible use in assays.
  • Provided in multiple package sizes suitable for research workflows.
  • High reported purity and defined molecular weight for analytical consistency.
Catalog No. 50-002-49119 Supplier Medchemexpress LLC Supplier No. HY101954A50MG
Inarigivir ammonium is a dinucleotide research compound that functions as a RIG-I agonist and has been reported to reduce hepatitis B virus (HBV) DNA in preclinical models. It is provided for laboratory research in solid form and as DMSO solutions, and is characterized by high reported purity and defined physicochemical data.
  • Acts as a RIG-I agonist to activate innate immune responses.
  • Demonstrated antiviral activity against hepatitis B virus in preclinical studies.
  • Supplied as a solid and as 10 mM DMSO solutions for flexible use in assays.
  • Provided in multiple package sizes suitable for research workflows.
  • High reported purity and defined molecular weight for analytical consistency.