Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inarigivir ammonium | 2172788-92-8 | MFCD34578292 | 99.0% | 604.53 g/mol | C20H29N8O10PS | 10 MG
SDP

Catalog No. 5000248509
Encompass_Preferred

Inarigivir ammonium is a dinucleotide antiviral research compound shown to reduce HBV DNA and RNA in preclinical models. It is supplied as a white to light-yellow solid with high chemical purity for laboratory research applications.

  • Dinucleotide antiviral active against hepatitis B virus.
  • High purity: 99.04% (HPLC).
  • Molecular weight 604.53 g/mol and formula C20H29N8O10PS.
  • Solid form suitable for in vitro and preclinical studies.
  • Available in small research quantities and DMSO solution formats.

Catalog No. 50-002-48509 Supplier Medchemexpress LLC Supplier No. HY101954A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inarigivir ammonium is a dinucleotide antiviral research compound shown to reduce HBV DNA and RNA in preclinical models. It is supplied as a white to light-yellow solid with high chemical purity for laboratory research applications.

  • Dinucleotide antiviral active against hepatitis B virus.
  • High purity: 99.04% (HPLC).
  • Molecular weight 604.53 g/mol and formula C20H29N8O10PS.
  • Solid form suitable for in vitro and preclinical studies.
  • Available in small research quantities and DMSO solution formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.