Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Inarigivir (ammonium salt) | 2172788-92-8 | 99.0% | 604.53 | C20H29N8O10PS | 100 MG
SDP

Catalog No. 5000248493
Encompass_Preferred

Inarigivir ammonium is a dinucleotide antiviral research compound reported to inhibit hepatitis B virus (HBV). It has demonstrated in vivo activity reducing liver HBV DNA in animal models and is supplied for laboratory research use with accompanying technical documentation and safety information.

  • Acts as a dinucleotide antiviral and HBV inhibitor.
  • Demonstrated in vivo reduction of liver HBV DNA in animal studies.
  • High purity of 99.0% by manufacturer analysis.
  • Molecular formula C20H29N8O10PS and molecular weight 604.53.
  • Soluble in DMSO and water at up to 100 mg/mL; requires ultrasonic assistance.
  • Available in multiple pack sizes, including 100 MG.
  • Includes datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-48493 Supplier Medchemexpress LLC Supplier No. HY101954A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inarigivir ammonium is a dinucleotide antiviral research compound reported to inhibit hepatitis B virus (HBV). It has demonstrated in vivo activity reducing liver HBV DNA in animal models and is supplied for laboratory research use with accompanying technical documentation and safety information.

  • Acts as a dinucleotide antiviral and HBV inhibitor.
  • Demonstrated in vivo reduction of liver HBV DNA in animal studies.
  • High purity of 99.0% by manufacturer analysis.
  • Molecular formula C20H29N8O10PS and molecular weight 604.53.
  • Soluble in DMSO and water at up to 100 mg/mL; requires ultrasonic assistance.
  • Available in multiple pack sizes, including 100 MG.
  • Includes datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.