Learn More
Medchemexpress LLC HY-109035 10mg Medchemexpress, Inarigivir soproxil CAS:942123-43-5 Purity:>98%
SDP
Catalog No. 501968654
Encompass_Preferred
Medchemexpress, HY-109035 10mg Inarigivir soproxil CAS:942123-43-5 Inarigivir soproxil is an agonist of innate immunity and shows potent antiviral activity against resistant hepatitis C virus (HCV) variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8654 Supplier Medchemexpress LLC Supplier No. HY10903510MG
Medchemexpress, HY-109035 10mg Inarigivir soproxil CAS:942123-43-5 Inarigivir soproxil is an agonist of innate immunity and shows potent antiviral activity against resistant hepatitis C virus (HCV) variants, with EC50s of 2.2 and 1.0 μM for HCV 1a/1b in cells of genotype 1 HCV replicon systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.