Learn More
Apexbio Technology LLC Inauhzin 309271-94-1 10mg
SDP
Catalog No. 502240495
Encompass_Preferred
Inauhzin (CAS 309271-94-1) is a small molecule inhibitor of SIRT1 exhibiting an IC50 of 0 7 2 M It selectively impedes SIRT1-mediated deacetylase activity in a dose-dependent manner leading to increased acetylation of histone H3 (at K382 and K9) and p53 Inauhzin does not significantly inhibit SIRT2 SIRT3 or HDAC8 in fluorescence-based assays Through activation of p53 Inauhzin induces apoptosis and suppresses proliferation in various human cancer cell lines with IC50 values ranging from 3 2 to 85 4 M depending on the cell type In xenograft models it promotes p53-dependent apoptosis highlighting its utility for studying SIRT1 inhibition and p53-mediated tumor suppression in oncology research
This item is not returnable. View return policy
Catalog No. 50-224-0495 Supplier Apexbio Technology LLC Supplier No. B327210
Inauhzin (CAS 309271-94-1) is a small molecule inhibitor of SIRT1 exhibiting an IC50 of 0 7 2 M It selectively impedes SIRT1-mediated deacetylase activity in a dose-dependent manner leading to increased acetylation of histone H3 (at K382 and K9) and p53 Inauhzin does not significantly inhibit SIRT2 SIRT3 or HDAC8 in fluorescence-based assays Through activation of p53 Inauhzin induces apoptosis and suppresses proliferation in various human cancer cell lines with IC50 values ranging from 3 2 to 85 4 M depending on the cell type In xenograft models it promotes p53-dependent apoptosis highlighting its utility for studying SIRT1 inhibition and p53-mediated tumor suppression in oncology research