Learn More
Medchemexpress LLC HY-15869 5mg Medchemexpress, Inauhzin CAS:309271-94-1 Purity:>98%
SDP
Catalog No. 502081808
Encompass_Preferred
Medchemexpress, HY-15869 5mg Inauhzin CAS:309271-94-1 Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1808 Supplier Medchemexpress LLC Supplier No. HY158695MG
Medchemexpress, HY-15869 5mg Inauhzin CAS:309271-94-1 Inauhzin is a dual SirT1/IMPDH2 inhibitor, and acts as an activator p53, used in the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.