Learn More
Medchemexpress LLC Inaxaplin | 2446816-88-0 | 99.4% | 417.38 g/mol | C21H18F3N3O3 | 5 MG
SDP
Catalog No. 5000262149
Encompass_Preferred
Inaxaplin (VX-147) is a small-molecule apolipoprotein L1 (APOL1) function inhibitor used in research on APOL1-mediated kidney disease. Supplied for research use only, the compound is characterized for in vitro and translational studies and is available as a high-purity solid and as a DMSO solution for biochemical and cell-based assays.
  • Potent APOL1 inhibition suitable for mechanism-of-action studies.
  • Intended for research use only and not for clinical use.
  • Available as a high-purity solid and as a 10 mM DMSO solution.
  • High reported purity supports reproducible experimental results.
  • Suitable for biochemical, cellular, and translational research on kidney disease.
Catalog No. 50-002-62149 Supplier Medchemexpress LLC Supplier No. HY1328205MG
Inaxaplin (VX-147) is a small-molecule apolipoprotein L1 (APOL1) function inhibitor used in research on APOL1-mediated kidney disease. Supplied for research use only, the compound is characterized for in vitro and translational studies and is available as a high-purity solid and as a DMSO solution for biochemical and cell-based assays.
  • Potent APOL1 inhibition suitable for mechanism-of-action studies.
  • Intended for research use only and not for clinical use.
  • Available as a high-purity solid and as a 10 mM DMSO solution.
  • High reported purity supports reproducible experimental results.
  • Suitable for biochemical, cellular, and translational research on kidney disease.