Learn More
TARGETMOL CHEMICALS INC INAXAPLIN 5MG
SDP
Catalog No. 502580004
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Inaxaplin is an apolipoprotein L1 ( APOL1 ) function inhibitor (WO2020131807 compound 2). Inaxaplin can be used for the research of kidney disease. purity: 98%
Catalog No. 50-258-0004 Supplier TARGETMOL CHEMICALS INC Supplier No. T401085MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Inaxaplin is an apolipoprotein L1 ( APOL1 ) function inhibitor (WO2020131807 compound 2). Inaxaplin can be used for the research of kidney disease. purity: 98%