Learn More
Medchemexpress LLC Inaxaplin (VX-147) | 2446816-88-0 | 99.4% | C21H18F3N3O3 | 10MG
SDP
Catalog No. 5000258200
Encompass_Preferred
Inaxaplin (VX-147) is an orally active small-molecule inhibitor of apolipoprotein L1 (APOL1) channel function, developed for research into APOL1-mediated kidney disease. It is supplied for laboratory research as a high-purity solid or as a DMSO solution.
Catalog No. 50-002-58200 Supplier Medchemexpress LLC Supplier No. HY13282010MG
Inaxaplin (VX-147) is an orally active small-molecule inhibitor of apolipoprotein L1 (APOL1) channel function, developed for research into APOL1-mediated kidney disease. It is supplied for laboratory research as a high-purity solid or as a DMSO solution.