Learn More
Darwin Chambers Company Triple Door Heat Humidity Chamber 66 cu. ft.
SDP
Catalog No. 501893886
Encompass_Preferred
The HH084 is a 76.2 cubic ft triple door heated/humidified incubator. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.
Catalog No. 50-189-3886 Supplier Darwin Chambers Company Supplier No. HH084AA
Add to Cart
Add to Cart
Request a Quote
The HH084 is a 76.2 cubic ft triple door heated/humidified incubator. Temperature range is 5°C above ambient to 50°C degrees C. RH ambient to 90 percent. This unit comes w/ 9 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior & stainless steel floor w/ coved corners. Running the models below will save you up to 80% in electricity costs vs. a compressor unit from competitors.