Learn More
World Precision Instrument Incapacitance Meter for Mouse and Rat
SDP
Catalog No. NC1957138
Encompass
Test pain and inflammation in small animal hind limbs
The Incapacitance Meter uses a technique called dual channel weight averaging, which tests both hind limbs. This gives you a clean, stress-free correlation of the paw pressure test. Conduct control and testing of the animal at the same time. Place the animal in the holder with its hind limbs resting on the two weight-averaging platform pads. The controller records the average weight (grams) over the test period as the animal shifts its weight from each pad.
Catalog No. NC1957138 Supplier World Precision Instrument Supplier No. II600MR
Add to Cart
Add to Cart
Request a Quote
Test pain and inflammation in small animal hind limbs
The Incapacitance Meter uses a technique called dual channel weight averaging, which tests both hind limbs. This gives you a clean, stress-free correlation of the paw pressure test. Conduct control and testing of the animal at the same time. Place the animal in the holder with its hind limbs resting on the two weight-averaging platform pads. The controller records the average weight (grams) over the test period as the animal shifts its weight from each pad.