Learn More
Apexbio Technology LLC INCB-024360, 10mM (in 1mL DMSO). Cas: 1204669-58-8 MFCD: MFCD29075639. potent and selective inhibitor of IDO1
SDP
Supplier:  Apexbio Technology LLC B60365.1
Encompass_Preferred
INCB024360 is a potent and selective inhibitor of IDO1 with IC50 value of 10 nM. [1]IDO means indoleamine-pyrrole 2, 3-dioxygenase. IDO is an enzyme which is encoded by the IDO1 gene. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5371