Learn More
Medchemexpress LLC HY-15689 2g Medchemexpress, INCB 024360 Epacadostat CAS:1204669-58-8 Purity:98%
SDP
Catalog No. 501638379
Encompass_Preferred
Medchemexpress,HY-15689 2g INCB 024360 Epacadostat CAS:1204669-58-8 Formula:C11H13BrFN7O4S INCB 024360 is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8379 Supplier Medchemexpress LLC Supplier No. HY156892G
Medchemexpress,HY-15689 2g INCB 024360 Epacadostat CAS:1204669-58-8 Formula:C11H13BrFN7O4S INCB 024360 is a potent and selective indoleamine 2,3-dioxigenase 1 (IDO1) inhibitor with an IC50 of 71.8 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.