Learn More
Cayman Chemical INCB 057643 10mg
SDP
Catalog No. 502990848
Encompass_Preferred
A BET family protein inhibitor; inhibits binding of BRD2, BRD3, and BRD4 to an acetylated histone H4 peptide in vitro; decreases Myc levels in, and inhibits proliferation of, AML, DLBCL, and MM cells; reduces tumor growth in AML, DLBCL, and MM mouse xenograft models; reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model
Catalog No. 50-299-0848 Supplier Cayman Chemical Supplier No. 3145610 MG
A BET family protein inhibitor; inhibits binding of BRD2, BRD3, and BRD4 to an acetylated histone H4 peptide in vitro; decreases Myc levels in, and inhibits proliferation of, AML, DLBCL, and MM cells; reduces tumor growth in AML, DLBCL, and MM mouse xenograft models; reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model