Learn More
Cayman Chemical INCB 3284 (dimesylate), Purity: ≥95%, CAS: 887401-93-6, 50 mg
SDP
Catalog No. 502954627
Encompass_Preferred
A potent, selective, and orally bioavailable antagonist of MCP-1 binding to CCR2 (IC50 = 3.7 nM); inhibits chemotaxis in vitro (IC50 = 4.7 nM) and displays acceptable oral bioavailability in mice, rats, dogs, monkeys, and chimpanzees
Catalog No. 50-295-4627 Supplier Cayman Chemical Supplier No. 1196350 MG
A potent, selective, and orally bioavailable antagonist of MCP-1 binding to CCR2 (IC50 = 3.7 nM); inhibits chemotaxis in vitro (IC50 = 4.7 nM) and displays acceptable oral bioavailability in mice, rats, dogs, monkeys, and chimpanzees