Learn More
Apexbio Technology LLC INCB024360 analogue, 10mg. Cas: 914471-09-3 MFCD: MFCD18633286. potent and selective inhibitor of IDO1
SDP
Supplier:  Apexbio Technology LLC A349310
Encompass_Preferred
INCB024360 analogue is a potent and selective inhibitor of IDO1 with IC50 value of 67 nM. [1]IDO (indoleamine-pyrrole 2, 3-dioxygenase) is an enzyme which is encoded by the IDO1 gene. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4806