Learn More
Apexbio Technology LLC INCB024360 analogue 914471-09-3 5mg
SDP
Catalog No. 5000063147
Encompass_Preferred
INCB024360 analogue (CAS 914471-09-3) is a selective inhibitor of indoleamine 2 3-dioxygenase 1 (IDO1) an enzyme responsible for initiating tryptophan catabolism along the kynurenine pathway IDO1-mediated depletion of tryptophan in the tumor microenvironment can suppress T-cell proliferation and promote immune tolerance facilitating cancer immune evasion INCB024360 analogue inhibits human IDO1 with an IC50 of 67 nM and demonstrates minimal activity against the related enzyme TDO In vivo studies in mouse melanoma models reveal dose-dependent anti-tumor activity and significant reductions in kynurenine levels supporting its utility as a research tool to investigate IDO1-targeted immunotherapy strategies
Catalog No. 50-000-63147 Supplier Apexbio Technology LLC Supplier No. A34935
Add to Cart
Add to Cart
INCB024360 analogue (CAS 914471-09-3) is a selective inhibitor of indoleamine 2 3-dioxygenase 1 (IDO1) an enzyme responsible for initiating tryptophan catabolism along the kynurenine pathway IDO1-mediated depletion of tryptophan in the tumor microenvironment can suppress T-cell proliferation and promote immune tolerance facilitating cancer immune evasion INCB024360 analogue inhibits human IDO1 with an IC50 of 67 nM and demonstrates minimal activity against the related enzyme TDO In vivo studies in mouse melanoma models reveal dose-dependent anti-tumor activity and significant reductions in kynurenine levels supporting its utility as a research tool to investigate IDO1-targeted immunotherapy strategies