Learn More
Selleck Chemical LLC INCB024360 analogue 5mg 914471-09-3 INCB14943
SDP
Supplier:  Selleck Chemical LLC S75875MG
Encompass_Preferred
INCB024360 analogue (INCB14943) is a potent, competitive IDO1 (indoleamine-(2,3)-dioxygenase) inhibitor with IC50 of 67 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7997