Learn More
Cayman Chemical INCB024360 nalog 10mg
SDP
Catalog No. 502979464
Encompass_Preferred
A selective IDO1 inhibitor (IC50s = 67 and 19 nM for human IDO in enzymatic activity and HeLa cell assays, respectively); decreases kynurenine levels in plasma and dose-dependently reduces the growth of GM-CSF-secreting B16 tumor xenografts in mice at 25-75 mg/kg
Catalog No. 50-297-9464 Supplier Cayman Chemical Supplier No. 1826510 MG
A selective IDO1 inhibitor (IC50s = 67 and 19 nM for human IDO in enzymatic activity and HeLa cell assays, respectively); decreases kynurenine levels in plasma and dose-dependently reduces the growth of GM-CSF-secreting B16 tumor xenografts in mice at 25-75 mg/kg