Learn More
Medchemexpress LLC HY-101870B 1mg Medchemexpress, INCB053914 (phosphate) CAS:2088852-47-3 Purity:>98%
SDP
Catalog No. 501967904
Encompass_Preferred
Medchemexpress, HY-101870B 1mg INCB053914 (phosphate) CAS:2088852-47-3 INCB053914 phosphate is an inhibitor of Pim extracted from patent WO 2017044730 A1, compound 1; has an IC50 of less than 35 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7904 Supplier Medchemexpress LLC Supplier No. HY101870B1MG
Medchemexpress, HY-101870B 1mg INCB053914 (phosphate) CAS:2088852-47-3 INCB053914 phosphate is an inhibitor of Pim extracted from patent WO 2017044730 A1, compound 1; has an IC50 of less than 35 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.